missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-Ngamma-trityl-D-asparagine, 95%

Brand:  Alfa Aesar™ H62029.03

Code : NEW

Additional Details : CAS Number : 210529-01-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478618

  • £28.73 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Boc-Ngamma-trityl-D-asparagine
474.55
Boc-D-Asn(Trt)-OH; (S)-4-(Boc-amino)-4-oxo-2-(tritylamino)butyric acid
210529-01-4
MFCD00153300

Specifications

Nalpha-Boc-Ngamma-trityl-D-asparagine
Powder
474.55
Boc-D-Asn(Trt)-OH; (S)-4-(Boc-amino)-4-oxo-2-(tritylamino)butyric acid
1g
210529-01-4
95%
MFCD00153300
White to off-white
Videos
SDS