missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Boc-Nepsilon-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.
Solubility
Soluble in dimethylformamide. Insoluble in water.
Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.
Solubility
Soluble in dimethylformamide. Insoluble in water.
Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 93°C to 98°C |
Color | White |
Beilstein | 3576584 |
Solubility Information | Soluble in dimethylformamide. Insoluble in water. |
IUPAC Name | (2S)-6-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Formula Weight | 468.55 |
Percent Purity | 98% |
Quantity | 5 g |
Odor | Odorless |
Physical Form | Solid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'