missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Nepsilon-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals

Catalog Number p-7053586
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
84624-27-1
C26H32N2O6
468.55
MFCD00062032
JYEVQYFWINBXJU-ANBDAQEENA-N
boc-lys fmoc-oh, n-boc-n'-fmoc-l-lysine, n2-boc-n6-fmoc-l-lysine, n-boc-n-fmoc-l-lysine, boclys fmoc, n-epsilon-fmoc-alpha-t-boc-l-lysine, 2s-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, pubchem14939, n, a-boc-n, a-fmoc-l-lysine
10983524
CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
84624-27-1
C26H32N2O6
468.55
MFCD00062032
JYEVQYFWINBXJU-ANBDAQEENA-N
boc-lys fmoc-oh, n-boc-n'-fmoc-l-lysine, n2-boc-n6-fmoc-l-lysine, n-boc-n-fmoc-l-lysine, boclys fmoc, n-epsilon-fmoc-alpha-t-boc-l-lysine, 2s-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, pubchem14939, n, a-boc-n, a-fmoc-l-lysine
10983524
CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.

Solubility
Soluble in dimethylformamide. Insoluble in water.

Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 93°C to 98°C
Color White
Beilstein 3576584
Solubility Information Soluble in dimethylformamide. Insoluble in water.
IUPAC Name (2S)-6-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
Formula Weight 468.55
Percent Purity 98%
Quantity 5 g
Odor Odorless
Physical Form Solid
Chemical Name or Material Nalpha-Boc-Nepsilon-Fmoc-L-lysine
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'