missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-Nepsilon-Fmoc-L-lysine, 98%

Brand:  Alfa Aesar™ H61126.14

Code : NEW

Additional Details : CAS Number : 84624-27-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15477628

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Applications
Nalpha-Boc-Nepsilon-Fmoc-L-lysine is used as a pharmaceutical intermediate.

Solubility
Soluble in dimethylformamide. Insoluble in water.

Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

Nalpha-Boc-Nepsilon-Fmoc-L-lysine
468.55
3576584
84624-27-1
MFCD00062032
Boc-Lys(Fmoc)-OH

Specifications

Nalpha-Boc-Nepsilon-Fmoc-L-lysine
Solid
468.55
3576584
White
25g
84624-27-1
98%
MFCD00062032
Boc-Lys(Fmoc)-OH
93° to 98°C
Odorless
Videos