missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-Ndelta-xanthyl-L-glutamine, 98%

Brand:  Alfa Aesar™ H63545.14

Code : NEW

Additional Details : CAS Number : 55260-24-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15470289

  • £94.45 / 25g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Boc-Ndelta-xanthyl-L-glutamine
426.46
Boc-Gln(Xan)-OH; Nα-Boc-Nδ-(9-xanthenyl)-L-glutamine, 98%
55260-24-7
MFCD00066158

Specifications

Nalpha-Boc-Ndelta-xanthyl-L-glutamine
Powder
426.46
Boc-Gln(Xan)-OH; Nα-Boc-Nδ-(9-xanthenyl)-L-glutamine, 98%
25g
55260-24-7
98%
MFCD00066158
White
Videos
SDS