missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-L-ornithine, 95%, Thermo Scientific Chemicals

Catalog Number 15498608
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
21887-64-9
C10H20N2O4
232.28
MFCD00076970
AMPVNPYPOOQUJF-ZETCQYMHSA-N
boc-orn-oh, boc-l-ornithine, n, a-boc-l-ornithine, l-ornithine, n2-1,1-dimethylethoxy carbonyl, 2s-5-amino-2-tert-butoxycarbonyl amino pentanoic acid, n2-boc-l-ornithine, nalpha-boc-l-ornithine, pubchem18983, nalpha-boc-l-ornithine;
7018784
(2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)(C)OC(=O)NC(CCCN)C(=O)O
This item is not returnable. View return policy
21887-64-9
C10H20N2O4
232.28
MFCD00076970
AMPVNPYPOOQUJF-ZETCQYMHSA-N
boc-orn-oh, boc-l-ornithine, n, a-boc-l-ornithine, l-ornithine, n2-1,1-dimethylethoxy carbonyl, 2s-5-amino-2-tert-butoxycarbonyl amino pentanoic acid, n2-boc-l-ornithine, nalpha-boc-l-ornithine, pubchem18983, nalpha-boc-l-ornithine;
7018784
(2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CC(C)(C)OC(=O)NC(CCCN)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 207°C to 208°C
Color White
Beilstein 4313353
Formula Weight 232.28
Percent Purity 95%
Quantity 1 g
Physical Form Powder
Chemical Name or Material Nalpha-Boc-L-ornithine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'