missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-L-asparagine 4-nitrophenyl ester, 98%

Brand:  Alfa Aesar™ H63765.14

Code : NEW

Additional Details : CAS Number : 4587-33-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15420629

  • £108.12 / 25g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Boc-L-asparagine 4-nitrophenyl ester
353.33
1895460
4587-33-1
MFCD00056109
Boc-Asn-ONp

Specifications

Nalpha-Boc-L-asparagine 4-nitrophenyl ester
Solid
353.33
1895460
25g
4587-33-1
98%
MFCD00056109
Boc-Asn-ONp
Videos
SDS