missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%, Thermo Scientific Chemicals

Catalog Number p-7055110
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
146987-10-2
C31H38N4O6S
594.73
MFCD00270741
OFIBQNGDYNGUEZ-CVKYCUBCNA-N
fmoc-lys biotin-oh, fmoc-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamido hexanoic acid, fmoc biocytin, fmoc-biocytin, fmoc-lys biotin, n, a-fmoc-biocytin, fmoc-l-lys biotinyl-oh
11039242
[H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2
This item is not returnable. View return policy
146987-10-2
C31H38N4O6S
594.73
MFCD00270741
OFIBQNGDYNGUEZ-CVKYCUBCNA-N
fmoc-lys biotin-oh, fmoc-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamido hexanoic acid, fmoc biocytin, fmoc-biocytin, fmoc-lys biotin, n, a-fmoc-biocytin, fmoc-l-lys biotinyl-oh
11039242
[H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2

Nα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.

Solubility
Slightly soluble in water and dimethyl sulfoxide.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Solubility Information Slightly soluble in water and dimethyl sulfoxide.
IUPAC Name (2S)-6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
Formula Weight 594.72
Percent Purity 95%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'