missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%

Brand:  Alfa Aesar™ H63609.06

Code : NEW

Additional Details : CAS Number : 146987-10-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15460389

  • £615.00 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Description

Description

Applications
Nα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.

Solubility
Slightly soluble in water and dimethyl sulfoxide.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine
594.72
Fmoc-Lys(biotinyl)-OH
146987-10-2
MFCD00270741

Specifications

Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine
Solid
594.72
Fmoc-Lys(biotinyl)-OH
146987-10-2
95%
MFCD00270741
5g
Videos
SDS