missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine dicyclohexylammonium salt, 95%

Brand:  Alfa Aesar™ H63222.03

Code : NEW

Additional Details : CAS Number : 16879-90-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478176

  • £10.07 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine dicyclohexylammonium salt
552.74
Z-Tyr(tBu)-OH.DCHA
16879-90-6
MFCD00038245

Specifications

Nalpha-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine dicyclohexylammonium salt
Solid
552.74
Z-Tyr(tBu)-OH.DCHA
16879-90-6
95%
MFCD00038245
1g
Videos
SDS