missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-Nomega-nitro-L-arginine, 98%, Thermo Scientific™

Catalog Number 15470059
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2304-98-5
C14H19N5O6
353.335
MFCD00038092
BZPCSFNCKORLQG-NSHDSACASA-N
z-arg no2-oh, z-l-arg no2-oh, n-cbz-n-nitro-l-arginine, cbz-l-arg no2-oh, s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid, l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl, cbz-arg no2-oh, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine, 2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, n-alpha-z-n-omega-nitro-l-arginine
12695023
C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
This item is not returnable. View return policy
2304-98-5
C14H19N5O6
353.335
MFCD00038092
BZPCSFNCKORLQG-NSHDSACASA-N
z-arg no2-oh, z-l-arg no2-oh, n-cbz-n-nitro-l-arginine, cbz-l-arg no2-oh, s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid, l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl, cbz-arg no2-oh, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine, 2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, n-alpha-z-n-omega-nitro-l-arginine
12695023
C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 353.33
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material Nalpha-Benzyloxycarbonyl-Nomega-nitro-L-arginine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'