missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-Ndelta-trityl-L-glutamine, 98%, Thermo Scientific™

Catalog Number 15420299
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
132388-60-4
C32H30N2O5
522.601
MFCD00144845
MYOAIKMOWHPBQS-NDEPHWFRSA-N
z-gln trt-oh, cbz-gln trt-oh, z-l-gln trt-oh, n-cbz-n'-trityl-l-glutamine, s-2-benzyloxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-benzyloxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem15365, 2s-2-benzyloxycarbonylamino-5-oxo-5-tritylamino pentanoic acid
10962399
(2S)-5-oxo-2-(phenylmethoxycarbonylamino)-5-(tritylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)NC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)O
This item is not returnable. View return policy
132388-60-4
C32H30N2O5
522.601
MFCD00144845
MYOAIKMOWHPBQS-NDEPHWFRSA-N
z-gln trt-oh, cbz-gln trt-oh, z-l-gln trt-oh, n-cbz-n'-trityl-l-glutamine, s-2-benzyloxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-benzyloxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem15365, 2s-2-benzyloxycarbonylamino-5-oxo-5-tritylamino pentanoic acid
10962399
(2S)-5-oxo-2-(phenylmethoxycarbonylamino)-5-(tritylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)NC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 522.59
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material Nalpha-Benzyloxycarbonyl-Ndelta-trityl-L-glutamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'