missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-L-alanine N-succinimidyl ester, 97%

Brand:  Alfa Aesar™ H63112.06

UNSPSC : 12352115

Code : NEW

Additional Details : CAS Number : 3401-36-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15498126

  • £28.83 / 5g


Estimated Shipment date
from Supplier 12-06-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Nalpha-Benzyloxycarbonyl-L-alanine N-succinimidyl ester
320.3
766066
3401-36-3
MFCD00053548
Z-Ala-OSu

Specifications

Nalpha-Benzyloxycarbonyl-L-alanine N-succinimidyl ester
Solid
320.3
766066
119° to 123°C
Moisture Sensitive
3401-36-3
97%
MFCD00053548
Z-Ala-OSu
5g
Documentation

Technical Documents