Learn More
Alfa Aesar™ Nalpha-Benzyloxycarbonyl-L-alanine N-succinimidyl ester, 97%
Brand: Alfa Aesar™ H63112.14
Code : NEW
Additional Details : CAS Number : 3401-36-3 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
Nalpha-Benzyloxycarbonyl-L-alanine N-succinimidyl ester | |
320.3 | |
766066 |
3401-36-3 | |
MFCD00053548 | |
Z-Ala-OSu |
Specifications
Nalpha-Benzyloxycarbonyl-L-alanine N-succinimidyl ester | |
Solid | |
320.3 | |
766066 | |
119° to 123°C | |
Moisture Sensitive |
3401-36-3 | |
97% | |
MFCD00053548 | |
Z-Ala-OSu | |
25g |