missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-D-2,3-diaminopropionic acid, 98%

Brand:  Alfa Aesar™ H61991.03

Code : NEW

Additional Details : CAS Number : 62234-37-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486906

  • £60.21 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Nalpha-Benzyloxycarbonyl-D-2,3-diaminopropionic acid
C11H14N2O4
MFCD00237346
Z-D-Dap-OH; (R)-3-Amino-2-(benzyloxycarbonylamino)propionic acid
62234-37-1
238.24
1993024

Specifications

Nalpha-Benzyloxycarbonyl-D-2,3-diaminopropionic acid
≈240°C (dec.)
62234-37-1
C11H14N2O4
MFCD00237346
Z-D-Dap-OH; (R)-3-Amino-2-(benzyloxycarbonylamino)propionic acid
+39° (c=1 in 1M HCl)
1g
98%
238.24
1993024
Videos
SDS