missing translation for 'onlineSavingsMsg'
Learn More

Naloxonazine dihydrochloride, Tocris Bioscience™

Selective μ1 antagonist

Brand:  Tocris (Bio-Techne) 059110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13427793

  • £119.07 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent opioid antagonist, selective for μ1 receptors.
Specifications

Chemical Identifiers

880759-65-9
723.692
naloxonazine dihydrochloride, bis-5-?-4,5-epoxy-3,14-dihydroxy-17-2-propenyl-morphinan-6-ylidene hydrazine dihydrochloride
(7E)-7-[(Z)-(4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-ylidene)hydrazinylidene]-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-4a,9-diol;dihydrochloride
C38H44Cl2N4O6
VIAIHLLKDJKEKM-QZOKRTFASA-N
131954536
C=CCN1CCC23C4C(=NN=C5CCC6(C7CC8=C9C6(C5OC9=C(C=C8)O)CCN7CC=C)O)CCC2(C1CC1=C3C(=C(C=C1)O)O4)O.Cl.Cl

Specifications

Naloxonazine dihydrochloride
880759-65-9
naloxonazine dihydrochloride, bis-5-?-4,5-epoxy-3,14-dihydroxy-17-2-propenyl-morphinan-6-ylidene hydrazine dihydrochloride
C=CCN1CCC23C4C(=NN=C5CCC6(C7CC8=C9C6(C5OC9=C(C=C8)O)CCN7CC=C)O)CCC2(C1CC1=C3C(=C(C=C1)O)O4)O.Cl.Cl
723.692
723.69
10mg
C38H44Cl2N4O6
VIAIHLLKDJKEKM-QZOKRTFASA-N
(7E)-7-[(Z)-(4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-ylidene)hydrazinylidene]-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-4a,9-diol;dihydrochloride
131954536
>98%
Videos
Documents