Learn More
Thermo Scientific™ Nalgene™ Stainless-Steel Forceps
Nalgene Stainless-Steel Forceps are available in bent or straight-tip styles. Bent-tip style is designed for easy removal and manipulation of filter membranes.
£38.55 - £43.15
Specifications
No. per Pack 1
View More Specs
Products 2
Product Code Brand Tip Style Price Quantity & Availability  
Product Code Brand Tip Style Price Quantity & Availability  
10010431 View Documents Thermo Scientific™
DS0399-0001
Bent £43.15
Each View alternative products
Save Up to 
»
Estimated Shipment
09-01-2020
Add to basket
10112161 View Documents Thermo Scientific™
DS0399-0002
Straight £38.55
Each View alternative products
Save Up to 
»
Estimated Shipment
09-01-2020
Description
Description
  • Stainless steel
  • Self-closing (bent tip)
  • Serrated gripping area
  • Autoclavable
Warranty: 90 days
Specifications
Specifications
1
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.