Learn More
Thermo Scientific™ Nalgene™ Stainless-Steel Forceps, straight tip
Nalgene Stainless-Steel Forceps are available in bent or straight-tip styles. Bent-tip style is designed for easy removal and manipulation of filter membranes.
Brand:  Thermo Scientific™ DS0399-0002
Code : A0
Additional Details : Weight : 0.05000kg
Product Code. 10112161
  • £37.17 / Each
Estimated Shipment: 19-08-2024
to see stock.
Add to basket
Description
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
  • Stainless steel
  • Self-closing (bent tip)
  • Serrated gripping area
  • Autoclavable
Specifications
Specifications
Straight, Flat, Serrated
Stainless Steel
Forceps
Ea.
1
Forceps
Stainless-Steel Forceps, straight tip
Straight