Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with SFCA Membrane
Perform vacuum filtration and single-use biological work requiring sterile filtration, prefiltration or clarification of aqueous solutions in the lab or field.
£77.34 - £345.99
Specifications
Membrane SFCA
No. per Pack 12
View More Specs
Products 14
Product Code Brand Volume (Metric) Upper Diameter (Metric) Membrane Pore Size Price Quantity & Availability  
Product Code Brand Volume (Metric) Upper Diameter (Metric) Membrane Pore Size Price Quantity & Availability  
10450241 View Documents Thermo Scientific™
122-0020
115mL 50mm 0.2μm £77.34
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
23-07-2020
Add to basket
10126070 View accessory products View Documents Thermo Scientific™
122-0045
115mL 50mm 0.45μm £77.34
Pack of 12 View alternative products
Save Up to 
»
In Stock 2
Add to basket
10454641 View accessory products View Documents Thermo Scientific™
155-0020
150mL 50mm 0.2μm £135.07
Pack of 12 View alternative products
Save Up to 
»
In Stock 24
Add to basket
10760792 View accessory products View Documents Thermo Scientific™
155-0045
150mL 50mm 0.45μm £93.77
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
23-07-2020
Add to basket
10444831 View accessory products View Documents Thermo Scientific™
157-0020
250mL 50mm 0.2μm £148.58
Pack of 12 View alternative products
Save Up to 
»
In Stock 4
Add to basket
10579051 View accessory products View Documents Thermo Scientific™
157-0045
250mL 50mm 0.45μm £104.07
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
23-07-2020
Add to basket
10017770 View accessory products View Documents Thermo Scientific™
156-4020
500mL 75mm 0.2μm £241.05
Pack of 12 View alternative products
Save Up to 
»
In Stock 19
Add to basket
10199700 View accessory products View Documents Thermo Scientific™
156-4045
500mL 75mm 0.45μm £244.17
Pack of 12 View alternative products
Save Up to 
»
In Stock 4
Add to basket
10211561 View accessory products View Documents Thermo Scientific™
162-0020
500mL 90mm 0.2μm £268.06
Pack of 12 View alternative products
Save Up to 
»
In Stock 3
Add to basket
10740602 View accessory products View Documents Thermo Scientific™
162-0045
500mL 90mm 0.45μm £258.71
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
23-07-2020
Add to basket
10614861 View Documents Thermo Scientific™
158-0045
1000mL 50mm 0.45μm £214.70
Pack of 12 View alternative products
Save Up to 
»
In Stock 1
Add to basket
10017820 View accessory products View Documents Thermo Scientific™
158-0020
1000mL 75mm 0.2μm £304.43
Pack of 12 View alternative products
Save Up to 
»
In Stock 3
Add to basket
10382711 View accessory products View Documents Thermo Scientific™
161-0020
1000mL 90mm 0.2μm £336.64
Pack of 12 View alternative products
Save Up to 
»
In Stock 11
Add to basket
10405021 View accessory products View Documents Thermo Scientific™
161-0045
1000mL 90mm 0.45μm £345.99
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
23-07-2020
Add to basket
Description
Description
Features
  • Upper chamber separates from storage bottle for easy access to filtrate
  • Upper chamber clearly marked with pore size, membrane type, catalog number, lot number and expiration date (except 115 mL size)
  • Leakproof† (except 115 mL sizes) screw cap eliminates pH shift in storage bottle to provide longer life for stored media
  • Padless membrane support minimizes foaming of proteinaceous samples
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
Surfactant-Free Cellulose Acetate Membranes (SFCA)
  • Low protein binding, excellent flow rates, little or no loss of specific proteins
  • No wetting agents to affect sensitive cell culture lines
  • Yellow collar for easy identification 
  • Much lower extractables than standard CA membranes 
115mL Sizes
  • Compact filter units for quick filtration of 50 to 100mL of fluid
  • Unitary construction with non-separable upper and lower compartments
  • Not intended for post-filtration storage of media (use Rapid-Flow products if media storage is required)
  • 115mL sizes do not have Rapid-Flow support plate design
Includes:
Polystyrene cover; graduated upper chamber and storage bottle (except 115mL size) integral membrane filter; cellulosic-plugged side arm; quick disconnect tubing adapter.
Warranty: 90 days
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Specifications
SFCA
12
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.