Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes
Increase filtration efficiency. PES membranes and exclusive Rapid-Flow design provide fast flow rates and high throughput while defending against cell culture contamination.
Brand: Thermo Scientific™ 150-0020
Code : 54
Additional Details : Weight : 1.66500kg
Description
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ Disposable Filter Units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid Flow Filter Units and Bottle Tops are the best choice for the filtration of cell culture media, serum, additives and buffers. Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see Full Specifications in the product table for item participation).
- Available in PES, CN, SFCA and Nylon membranes. Devices are color coded according to membrane.
- Devices feature the fastest flow rate and lowest clogging of comparable alternatives
- 0.1um PES available for mycoplasma protection
- 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
- Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit.
- Available with Fit for Purpose Sterile documentation
- Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see "Full Specifications" in the product table for item participation)
Specifications
Single-use | |
0.2 μm | |
12 | |
Filter Unit | |
150 mL | |
Sterile |
Nylon | |
Sterile | |
12 | |
150 mL | |
50 mm |