missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PPCO Low-Profile Closures for Micro Packaging Vials: Nonsterile, Bulk Pack

Identify samples quickly with low-profile, color-coded closures molded of high-purity, low metal content PPCO resin.

Brand:  Thermo Scientific™ 362821-1112

Code : NEW

Additional Details : Weight : 0.39000kg

 View more versions of this product

Product Code. 10099392

  • £99.82 / Pack of 1000
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

Quickly identify samples using Thermo Scientific™ Nalgene™ PPCO Low-Profile Micro Packaging Vial Closures, offered in a variety of colors. Closures are molded of high-purity, low metal-content polypropylene copolymer resin—excellent for PCR reagents.

Features:

  • Vials (listed separately) and closures are pressure-tested together at 7.5 PSIG for air shipment
  • Nonsterile
  • Noncytotoxic and nonpyrogenic
  • Leakproof† when used with Nalgene Micro Packaging Vials

Ordering Information:

Micro Packaging Vial Closures are bulk packed without vials, 500 per inner bag, 1,000 per case. Order nonsterile vials separately (Cat. Nos. 362800, 362805).

Specifications

Specifications

Low-profile closures for micro packaging vials
11 mm
Non-sterile
500
PPCO
ISO 13485:2016
11 mm
Pink
PPCO
1000
Nalgene
500 Bag, 1000 Cs.
Closure
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.