missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate, Validation Bottle

Catalog Number p-9078015
Click to view available options
Capacity (Metric):
30 mL
250 mL
2 L
This item is not returnable. View return policy
This item is not returnable. View return policy

For Use With: Perform validation testing; Autoclavable; Closure Material: Polypropylene; Color: White (Translucent)

Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.

Autoclavable, transparent polycarbonate construction allows for convenient material compatibility validation.

Includes:
White polypropylene screw closure and TPE gasket

TRUSTED_SUSTAINABILITY
Capacity (English) 1 oz.
Product Type Validation Bottle
Closure Material Polypropylene
Closure Size 20-415mm
Diameter (Metric) Inner Neck 14 mm
Diameter (Metric) Outer 32 mm
Capacity (Metric) 30 mL
Material Polycarbonate
For Use With (Application) Perform validation testing
Height (Metric) With Closure 75 mm
No. per Case 30
No. per Pack 30
Autoclavable Autoclavable
Sterility Non-sterile
Color White (Translucent)
Mouth Style Wide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'