Thermo Scientific™ Nalgene™ Polycarbonate Safety Splash Shield

Simultaneously maintain safety and a clear field of vision by using this safety splash shield. Provides protection from splashes, aerosols and flying debris.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: 6356-0001

X4 SAFETY SPLASH SHIELD

UNSPSC: 26142300

Code: Z4

Additional Details:
Additional Details: Weight: 2.93000kg



Alerts: Warning!: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.

Product Code. 11339554

Quantity Price
1 £ 407.94 / Pack of 4
Estimated Shipment
20-03-2017
Add to basket


Description and Specification

Specification

Item Description Shield Safety Splash PC 21.6 x 27.9 cm
Description Safety Splash Shield with acetal flexible arm
Lens Material Polycarbonate 
Certifications/Compliance Meets OSHA 29CFR1910.1030 standard
No. per Pack 1
No. per Case 4
Width (Metric) 21.5cm
Length (Metric) 28cm
Thickness (Metric) 3mm

  • Flexible 14 inch arm makes it easy to maneuver shield over samples
  • Magnetic base firmly grips to magnetic surfaces letting you position the shield horizontally (flat), vertically or on an overhead surface
  • Provides UV protection from 200-360nm for added versatility
  • Biohazard/Transparent
Warning!: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.