missing translation for 'onlineSavingsMsg'
Learn More

Nalgene™ Polycarbonate Fernbach Culture Flask

Increase aeration for better growth of culture in liquid media with the wide surface area of these Nalgene Fernbach culture flasks.

Brand:  Thermo Scientific Nalgene 4105-2800

Code : 54

Additional Details : Weight : 3.40000kg

Product Code. 10438612

  • £71.15 / Each


In Stock 4
Add to basket

Disclaimers:

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.


Description

Description

  • Break-resistant, transparent polycarbonate
  • Fernbach design supports better aeration
  • For leakproof seal† use No. 12 rubber stopper
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. B) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications

Specifications

2.8L capacity; D x H: 8 x 9 in.
2800mL
22.9cm
1
20.3cm
Polycarbonate
4
None
Videos