Learn More
Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
Description
Safely store and ship light-sensitive, diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque, amber, high-density polyethylene bottles are for use with light-sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
- Nonsterile
- Durable, uniform walls resistant to splitting or puncturing
- Leakproof† when assembled with closure (must be ordered separately)
Ordering Information:
Bottles are bulk packed without closures. Packed 2,000 per case. Order colored closures separately (Cat. No. 362150).
Spécification
Capacity | 0.25 oz., 8 mL |
Closure Material | Polypropylene |
Closure Size | 20-415 mm |
Diameter (Metric) | 25 mm |
Height (English) | 1.77 in. |
Height (Metric) | 45 mm |
Width (English) | 0.98 in. |
Product Type | Diagnostic Bottle |
Capacity (Metric) | 8 mL |
Capacity (English) | 0.25 oz. |
Afficher plus |
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.