missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™

Conduct high-speed centrifugation of aggressive chemicals with these tubes, suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.

Brand:  Thermo Scientific™ 3114-0050

95.40 GBP valid until 2024-04-26
Use promo code "21699" to get your promotional price.



Code : 55

Additional Details : Weight : 0.42480kg

 View more versions of this product

Product Code. 10384831

  • £168.00 / Pack of 2
In Stock
to see stock.
Add to basket

Alert:

Note: Must be filled to 100% of total capacity for centrifugation


Description

Description

Conduct high-speed centrifugation of aggressive chemicals with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™, specifically suited for chloroform and phenol extractions.

Features:

  • Teflon™ FEP, Tefzel™ ETFE screw closure
  • Withstand temperatures from -100° to +150°C (-148° to +302°F)
  • Leakproof† Tefzel ETFE closures prevent material loss under ordinary use for most spins <10,000 × g
  • Sealing caps available for 30mL and 50mL size to assure leakproof service especially at high speeds >10,000 × g or when spinning hazardous materials
  • Autoclavable/biohazard/leakproof†/PTFE FEP

Ordering Information:

Optional sealing caps available: DS3131-0020 (for 30mL tube) and DS3131-0024 (for 50mL tube). Cat. No. 3114-0050 has a brim capacity of only 42mL but is designed to fit standard rotor.

Includes:

Tefzel ETFE screw caps. Additional sealing cap assembly available separately (DS3131-series).

Specifications

Specifications

ETFE
Teflon FEP
2
Autoclavable
Round
No
No
50,000 x g (with sealing cap assembly)
50 mL
10
Non-sterile
Round
Screw Cap
28.8 mm
Centrifuge Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.