missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Economy Bottles

Catalog Number 10020741
Click to view available options
Capacity (Metric):
30 mL
125 mL
500 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Conduct light-duty, general purpose applications where a heavier bottle is not required with these narrow-mouth amber HDPE bottles.

Conduct light-duty, general purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Bottles. Lightweight versions of the original Nalgene Amber Lab Quality bottle, these high-density polyethylene bottles reduce UV light transmissions to protect light-sensitive liquids.

  • Meets U.S. Pharmacopoeia 23 requirements for light transmission
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted
  • Economical and recyclable; designed for single use
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

TRUSTED_SUSTAINABILITY
Capacity 1 oz., 30mL
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 20 mm
Closure Type Screw Cap
Diameter (Metric) 34 mm
Height (English) 2.4 in.
Height (Metric) 61 mm
Through the Closure Dispensing No
Width (English) 1.35 in.
Product Type Narrow Mouth Bottle
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Opaque Amber
Description Amber Narrow-Mouth Economy Bottle
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 72
No. per Pack 72
Sterility Non-sterile
Closure Color Amber
Neck Finish 20mm
Certifications/Compliance Meets the requirements for light-resistant containers per USP latest edition
Closure Included Yes
Disposable Single-use
Graduated No
Mouth Narrow
Shape Round
Show More Show Less

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'