missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure

Prevent permeation loss of samples with the Thermo Scientific™ Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure.

Brand:  Thermo Scientific™ 2124-0005

Additional Details : Weight : 1.06000kg

Product Code. 11379654

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Fluorinated to enhance long-term performance and prevent solvent permeation loss
  • Wide mouth allows access
  • Yields lower extractables than other bottles
  • NOTE: This bottle is not liquid leakproof
Includes: 
  • Fluorinated polypropylene screw closures

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications

Specifications

2L
89mm
6
Nonsterile
120mm
Wide
Wide-Mouth Bottle
Fluorinated HDPE
1
100mm
244mm
Videos
Alternative Products