Learn More
Thermo Scientific™ Cryo Tube Holders
Simplify processes with these benchtop tube holders for single-handed capping and decapping.
Brand:  Thermo Scientific™ 5030-0505
Code : 55
Additional Details : Weight : 0.67000kg
 View more versions of this product
Product Code. 12757045
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
Nalgene™ Vial Holder, 5x5 staggered compartments
5 x 5
1
Polycarbonate
Nalgene™
Yes
Vial Holder, 5x5 staggered compartments
4
Cryogenic tube holder
Videos
Documentation View accessory products