Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Barbed Bulkhead Fittings, 13 mm
For Use With: Nalgene Bottle and Carboy Closures
Brand: Thermo Scientific™ 6149-0001
Code : 55
Additional Details : Weight : 0.02340kg
Description
Customize tubing sets as culture vessels for aseptic transfer of liquids with autoclavable Thermo Scientific™ Nalgene™ Barbed Polypropylene Bulkhead Fittings, suitable for use with most Nalgene bottle and carboy closures. Pre-drilled closures allow easy, safe in-house assembly.
Features:- Closures accept (two) 0.25″ fittings
- Fittings, barbed at both ends, accept tubing for inside and outside container
Includes:
Instructions, drilling template and two each of polypropylene barbed fitting, acetyl nuts, silicone gaskets and and TPE port caps.
Specifications
Fittings | |
13 mm | |
Nalgene bottles and carboys |
0.5 in. | |
Polypropylene |