Learn More
Thermo Scientific™ Nalgene™ Syringe Filters, Non-sterile
Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
Brand: Thermo Scientific™ 727-2045
204.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.22500kg
Description
Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.
Key Features- 10 to 100mL sample size
- Inlet: Female Luer-Lok™
- Outlet: Male Luer slip
- Polypropylene housing with ID color rings
- Pore size and membrane type printed on unit
Surfactant-free Cellulose Acetate (SFCA)
- Low protein binding, lower extractables than standard CA
- Good for cell culture fluids and cellular solutions
- Polypropylene housings with blue color rings
Cellulose Acetate (CA)
- Some protein binding and extractables
- For sterile filtration and clarification of: buffers, aqueous solutions and reagents
- Polypropylene housings with red color rings
Nylon
- Naturally hydrophilic, low extractables, some protein binding
- Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
- Polypropylene housings with pink color rings
- Naturally hydrophobic
- Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
- Polypropylene housings with white color rings
Specifications
Nylon | |
Non-sterile | |
Fewer resources used and less waste | |
Nalgene™ | |
10 to 100 mL (Sample) | |
25 mm |
0.45 μm | |
Filtration of Cryprotectants such as DMSO, aqueous biological solutions, alcoholic solutions and nonaqueous/HPLC/GC solvent applications | |
Polypropylene (Housing) | |
Syringe Filter | |
25 mm |