Learn More
Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Vacuum Tubing
Make practical and efficient use of this 180 clear plastic vacuum tubing, thick-walled PVC, which can withstand a full vacuum (29.9 in. (760mm) Hg) at room temperature. Material is food-grade and complies with USP VI and USDA regulations.
POA
Specifications
Certifications/Compliance | Material is Food-Grade and complies with USP VI and USDA regulations. |
---|---|
Material | PVC |
Description
Make practical and efficient use of Thermo Scientific™ Nalgene™ 180 Clear Plastic Vacuum Tubing, thick-walled PVC, which can withstand a full vacuum (29.9 in. (760mm) Hg) at room temperature. Material is food-grade and complies with USP VI and USDA regulations.
Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com.Specifications
Material is Food-Grade and complies with USP VI and USDA regulations. | |
PVC |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Note:
Not intended for medical use. Nalgene 180 and 980 PVC tubing contain DEHP [Bis (2-ethylhexyl)] phthalate, a commonly used plasticizer. It is known to the state of California Environmental Agency that this chemical causes cancer or reproductive toxicity.