missing translation for 'onlineSavingsMsg'
Learn More

Nafamostat mesylate, Tocris Bioscience™

Serine protease inhibitor, inhibits tryptase

Brand:  Tocris (Bio-Techne) 308110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462354

  • £95.85 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Broad spectrum serine protease inhibitor. Displays selectivity for human tryptase when used at relatively low concentrations. Reduces eosinophil infiltration, mast cell activation and airway responsiveness in a murine model of asthma.
Specifications

Chemical Identifiers

82956-11-4
539.578
nafamostat mesylate, futhan, nafamostat mesilate, nafamostat mesylate, nafamstat mesilate, 6-amidino-2-naphthyl 4-guanidino-benzoate dimethanesulphonate, 6-amidino-2-naphthyl-4-guanidinobenzoate dimethanesulfonate, nafamostat mesylate fut-175, nafamstatmesilate, ronastat tn
(6-carbamimidoylnaphthalen-2-yl) 4-(diaminomethylideneamino)benzoate;methanesulfonic acid
C21H25N5O8S2
SRXKIZXIRHMPFW-UHFFFAOYSA-N
5311180
CS(=O)(=O)O.CS(=O)(=O)O.C1=CC(=CC=C1C(=O)OC2=CC3=C(C=C2)C=C(C=C3)C(=N)N)N=C(N)N

Specifications

Nafamostat mesylate
82956-11-4
nafamostat mesylate, futhan, nafamostat mesilate, nafamostat mesylate, nafamstat mesilate, 6-amidino-2-naphthyl 4-guanidino-benzoate dimethanesulphonate, 6-amidino-2-naphthyl-4-guanidinobenzoate dimethanesulfonate, nafamostat mesylate fut-175, nafamstatmesilate, ronastat tn
CS(=O)(=O)O.CS(=O)(=O)O.C1=CC(=CC=C1C(=O)OC2=CC3=C(C=C2)C=C(C=C3)C(=N)N)N=C(N)N
539.578
539.59
10mg
C21H25N5O8S2
SRXKIZXIRHMPFW-UHFFFAOYSA-N
(6-carbamimidoylnaphthalen-2-yl) 4-(diaminomethylideneamino)benzoate;methanesulfonic acid
5311180
>99%
Videos