missing translation for 'onlineSavingsMsg'
Learn More

Nafadotride, Tocris Bioscience™

Highly potent, preferential D3 antagonist

Brand:  Tocris (Bio-Techne) 134710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13439203

  • £149.85 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly potent, competitive, preferential dopamine D3 receptor antagonist. Ki values are 0.52, 5, and 269 nM for human cloned D3, D2 and D4 receptors respectively. Centrally active upon systemic administration.
Specifications

Chemical Identifiers

149649-22-9
365.477
nafadotride, +--nafadotride, n-1-butylpyrrolidin-2-yl methyl-4-cyano-1-methoxynaphthalene-2-carboxamide, n-1-butyl-2-pyrrolidinyl methyl-4-cyano-1-methoxy-2-naphthalenecarboxamide, d0w1hm, gtpl44, 4-cyano-1-methoxy-naphthalene-2-carboxylic acid 1-butyl-pyrrolidin-2-ylmethyl-amide, n-1-butyl-2-pyrrolidinyl methyl-4-cyano-1-methoxy-2naphthalenecarboxamide
CHEBI:64191
CCCCN1CCCC1CNC(=O)C2=C(C3=CC=CC=C3C(=C2)C#N)OC
C22H27N3O2
IDZASIQMRGPBCQ-UHFFFAOYSA-N
3408722
N-[(1-butylpyrrolidin-2-yl)methyl]-4-cyano-1-methoxynaphthalene-2-carboxamide

Specifications

Nafadotride
149649-22-9
nafadotride, +--nafadotride, n-1-butylpyrrolidin-2-yl methyl-4-cyano-1-methoxynaphthalene-2-carboxamide, n-1-butyl-2-pyrrolidinyl methyl-4-cyano-1-methoxy-2-naphthalenecarboxamide, d0w1hm, gtpl44, 4-cyano-1-methoxy-naphthalene-2-carboxylic acid 1-butyl-pyrrolidin-2-ylmethyl-amide, n-1-butyl-2-pyrrolidinyl methyl-4-cyano-1-methoxy-2naphthalenecarboxamide
CCCCN1CCCC1CNC(=O)C2=C(C3=CC=CC=C3C(=C2)C#N)OC
365.477
CHEBI:64191
>98%
10mg
C22H27N3O2
IDZASIQMRGPBCQ-UHFFFAOYSA-N
N-[(1-butylpyrrolidin-2-yl)methyl]-4-cyano-1-methoxynaphthalene-2-carboxamide
3408722
365.47
Videos