missing translation for 'onlineSavingsMsg'
Learn More

NAD 299 hydrochloride, Tocris Bioscience™

Selective, high affinity 5-HT1A antagonist

Brand:  Tocris (Bio-Techne) 328250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13432714

  • £643.86 / 50mg

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective, high affinity 5-HT1A receptor antagonist (Ki = 0.6 nM in vitro). Enhances the action of selective 5-HT reuptake inhibitors and reverses citalopram -induced (Cat. No. 1427) inhibition of serotonergic cell firing.
Specifications

Chemical Identifiers

184674-99-5
354.85
nad 299 hydrochloride, 3r-3-dicyclobutylamino-8-fluoro-3,4-dihydro-2h-1-benzopyran-5-carboxamide hydrochloride
(3R)-3-[di(cyclobutyl)amino]-8-fluoro-3,4-dihydro-2H-chromene-5-carboxamide;hydrochloride
C18H24ClFN2O2
GSZJANKLCPHEEX-BTQNPOSSSA-N
90488823
C1CC(C1)N(C2CCC2)C3CC4=C(C=CC(=C4OC3)F)C(=O)N.Cl

Specifications

50mg
184674-99-5
nad 299 hydrochloride, 3r-3-dicyclobutylamino-8-fluoro-3,4-dihydro-2h-1-benzopyran-5-carboxamide hydrochloride
C1CC(C1)N(C2CCC2)C3CC4=C(C=CC(=C4OC3)F)C(=O)N.Cl
354.85
354.85
NAD 299 hydrochloride
C18H24ClFN2O2
GSZJANKLCPHEEX-BTQNPOSSSA-N
(3R)-3-[di(cyclobutyl)amino]-8-fluoro-3,4-dihydro-2H-chromene-5-carboxamide;hydrochloride
90488823
>98%
Videos
Documents