Learn More
Macherey-Nagel™ N9 Polypropylene Transparent Screw Caps
Seal vials with screw caps with a secure septum to receive needles. The Macherey-NagelTM N9 pollypropylene transparent screw caps come with a choice of septa.
Brand:  Macherey-Nagel™ 702288
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10535761
  • £27.55 / Pack of 100
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • Screw closures N 9
  • Large range of screw closures with different septum materials and cap colors
  • Bonded screw caps (septa firmly connected with the cap; cannot be removed) – advantage: thick (blunt) HPLC needles cannot push the septum into the vial
  • Polymagnetic screw closure 702166 for CTC GC PAL
Specifications
Specifications
Transparent
Silicone white/PTFE blue, slitted
N9
Polypropylene, PTFE, Silicone
1 mm