Learn More
Tocris Bioscience™ N20C hydrochloride
Non-competitive NMDA open-channel blocker
Brand:  Tocris Bioscience™ 221310
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13440814
  • £106.65 / 10mg
Estimated Shipment
01-07-2019
Description
Description
Selective, non-competitive NMDA receptor antagonist (IC50 = 5 μM); binds to the receptor-associated ion channel and prevents glutamate-induced Ca2+ influx. Displays neuroprotective activity in vivo. Brain-penetrant.
Specifications
Specifications
N20C hydrochloride
>98
10mg
N20C hydrochloride
304.82