missing translation for 'onlineSavingsMsg'
Learn More

N1-[4-(Trifluoromethoxy)phenyl]-2-chloroacetamide, 97%, Thermo Scientific™

Catalog Number p-3486347
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
161290-85-3
C9H7ClF3NO2
253.61
MFCD00219796
MIWYZHHKNDZBCB-UHFFFAOYSA-N
2-chloro-n-4-trifluoromethoxy phenyl acetamide, n-chloroacetyl-4-trifluoromethoxy aniline, n1-4-trifluoromethoxy phenyl-2-chloroacetamide, buttpark 30\07-62, acetamide,2-chloro-n-4-trifluoromethoxy phenyl, 2-chloro-4'-trifluoromethoxy acetanilide, pubchem8402, timtec-bb sbb001832
601031
2-chloro-N-[4-(trifluoromethoxy)phenyl]acetamide
FC(F)(F)OC1=CC=C(NC(=O)CCl)C=C1
This item is not returnable. View return policy
161290-85-3
C9H7ClF3NO2
253.61
MFCD00219796
MIWYZHHKNDZBCB-UHFFFAOYSA-N
2-chloro-n-4-trifluoromethoxy phenyl acetamide, n-chloroacetyl-4-trifluoromethoxy aniline, n1-4-trifluoromethoxy phenyl-2-chloroacetamide, buttpark 30\07-62, acetamide,2-chloro-n-4-trifluoromethoxy phenyl, 2-chloro-4'-trifluoromethoxy acetanilide, pubchem8402, timtec-bb sbb001832
601031
2-chloro-N-[4-(trifluoromethoxy)phenyl]acetamide
FC(F)(F)OC1=CC=C(NC(=O)CCl)C=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 253.61
Percent Purity 97%
Chemical Name or Material N1-[4-(Trifluoromethoxy)phenyl]-2-chloroacetamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'