missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-pyridyl)benzamide, Thermo Scientific™

Catalog Number p-4187342
Click to view available options
Quantity:
10 g
25 g
50 g
This item is not returnable. View return policy
5221-44-3
C12H10N2O
198.225
MFCD00160355
PNWVOLKZHJEWQU-UHFFFAOYSA-N
n-pyridin-4-yl benzamide, 4-benzamidopyridine, n1-4-pyridyl benzamide, n-4-pyridylbenzamide, 4-benzoylaminopyridine, pyridine, 4-benzamido, n-4-pyridyl benzamide, benzamide, n-4-pyridinyl-9ci, n-benzoyl-4-aminopyridine, phenyl-n-4-pyridyl carboxamide
78893
N-pyridin-4-ylbenzamide
C1=CC=C(C=C1)C(=O)NC2=CC=NC=C2
This item is not returnable. View return policy
5221-44-3
C12H10N2O
198.225
MFCD00160355
PNWVOLKZHJEWQU-UHFFFAOYSA-N
n-pyridin-4-yl benzamide, 4-benzamidopyridine, n1-4-pyridyl benzamide, n-4-pyridylbenzamide, 4-benzoylaminopyridine, pyridine, 4-benzamido, n-4-pyridyl benzamide, benzamide, n-4-pyridinyl-9ci, n-benzoyl-4-aminopyridine, phenyl-n-4-pyridyl carboxamide
78893
N-pyridin-4-ylbenzamide
C1=CC=C(C=C1)C(=O)NC2=CC=NC=C2
CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 198.22
Percent Purity 97%
Chemical Name or Material N1-(4-Pyridyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'