missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Butylphenyl)acetamide, 97%, Thermo Scientific™

Catalog Number 10253242
Click to view available options
Amber Glass Bottle; 10g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 3663-20-5
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H17NO
Linear Formula CH3CH2CH2CH2C6H4NHC(O)CH3
MDL Number MFCD00277211
Quantity 10 g
Synonym n-4-butylphenyl acetamide, 4-butylacetanilide, n1-4-butylphenyl acetamide, acetamide, n-4-butylphenyl, gnf-pf-1449, p-n-butylacetanilid, 4'-butylacetoanilide, p-normalbutylacetanilide, maybridge1_000046, n-acetyl-4-n-butylaniline
InChI Key GDQKURDANTWHBA-UHFFFAOYSA-N
SMILES CCCCC1=CC=C(C=C1)NC(=O)C
IUPAC Name N-(4-butylphenyl)acetamide
Molecular Weight (g/mol) 191.274
PubChem CID 77227
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material N1-(4-Butylphenyl)acetamide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'