missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N1-(2-Chlorophenyl)-2-bromoacetamide, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 95% |
Linear Formula | ClC6H4NHC(O)CH2Br |
Quantity | 1 g |
Formula Weight | 248.51 |
Percent Purity | 95% |
Chemical Name or Material | N1-(2-chlorophenyl)-2-bromoacetamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'