missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Vinylcarbazole, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 63°C to 65°C |
Color | White to Yellow |
Boiling Point | 154°C (3.0 mmHg) |
Flash Point | 182°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Quantity | 5 g |
Show More |
Safety and Handling
Product Identifier
- N-Vinylcarbazole
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'