missing translation for 'onlineSavingsMsg'
Learn More

n-Undecane, 99%, Thermo Scientific Chemicals

Catalog Number p-7049945
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
1120-21-4
C11H24
156.31
MFCD00008959
RSJKGSCJYJTIGS-UHFFFAOYSA-N
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
14257
CHEBI:46342
undecane
CCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
1120-21-4
C11H24
156.31
MFCD00008959
RSJKGSCJYJTIGS-UHFFFAOYSA-N
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
14257
CHEBI:46342
undecane
CCCCCCCCCCC

n-Undecane is used as a mild sex attractant for various types of moths and cockroaches. It is also used as an internal standard in the gas chromatographic analysis. Further, it is used as lubricants and lubricant additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -26°C
Color Colorless
Density 0.74 g/mL
Boiling Point 196°C
Flash Point 60°C (140°F)
Odor Mild
Linear Formula CH3(CH2)9CH3
Refractive Index 1.4173
Quantity 50 g
UN Number UN2330
Solubility Information Miscible with ethyl alcohol and ether. Immiscible with water.
Formula Weight 156.3
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Undecane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'