missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[(Trimethylsilyl)methyl]benzylamine, 95%

Brand:  Alfa Aesar™ H64450.14

Code : NEW

Additional Details : CAS Number : 53215-95-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15431669

  • £68.20 / 25g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

N-[(Trimethylsilyl)methyl]benzylamine
193.36
2802488
53215-95-5
MFCD00012192
N-Benzyl-1-(trimethylsilyl)methylamine

Specifications

N-[(Trimethylsilyl)methyl]benzylamine
89° to 90°C (5mmHg)
25g
1.4960
95%
MFCD00012192
N-Benzyl-1-(trimethylsilyl)methylamine
0.88g/mL
Pale yellow
53215-95-5
Liquid
193.36
2802488
Videos
SDS