missing translation for 'onlineSavingsMsg'
Learn More

n-Tridecanoic Acid 97%, ACROS Organics™

Brand:  Acros Organics 139520500

Code : 29

Additional Details : CAS Number : 638-53-9 Weight : 0.10000kg

Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10550584

  • £124.60 / 50g


Estimated Shipment
11-06-2018
Add to basket

Specifications

Chemical Identifiers

n-Tridecanoic acid
638-53-9
100.0
97%
CH3(CH2)11CO2H
MFCD00002741
97%
96.0
≥96.0%
C13H26O2
214.35

Specifications

n-Tridecanoic acid
97%
96.0
Crystalline Chunks, Flakes or Powder
97%
CH3(CH2)11CO2H
214.35
02, 364
40° to 45°C
50g
638-53-9
100.0
≥96.0%
C13H26O2
Authentic
MFCD00002741
Glass bottle
White to pale yellow
Videos
SDS