missing translation for 'onlineSavingsMsg'
Learn More

n-Tridecane, 99+%, Thermo Scientific Chemicals

Catalog Number p-3737622
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
629-50-5
C13H28
184.37
MFCD00008979
IIYFAKIEWZDVMP-UHFFFAOYSA-N
n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group
12388
CHEBI:35998
tridecane
CCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
629-50-5
C13H28
184.37
MFCD00008979
IIYFAKIEWZDVMP-UHFFFAOYSA-N
n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group
12388
CHEBI:35998
tridecane
CCCCCCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -5°C to 4°C
Color Colorless
Density 0.7500g/mL
Boiling Point 234.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11CH3
Refractive Index 1.4240 to 1.4260
Quantity 25 mL
Beilstein 01, 171
Specific Gravity 0.75
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol
Formula Weight 184.36
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Tridecane
Show More Show Less
compliance-icons
Product Identifier
  • n-Tridecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'