missing translation for 'onlineSavingsMsg'
Learn More

n-Tetracontane, 97% min, Thermo Scientific Chemicals

Catalog Number p-7048698
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
4181-95-7
C40H82
563.10
MFCD00015267
KUPLEGDPSCCPJI-UHFFFAOYSA-N
n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc
20149
tetracontane
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
4181-95-7
C40H82
563.10
MFCD00015267
KUPLEGDPSCCPJI-UHFFFAOYSA-N
n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc
20149
tetracontane
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

n-Tetracontane is used in a passivation layer in copper-phthalocyanine field-effect transistors for improving the charge carrier mobility and used to modify the quasi two-dimensional surface electron system on Au(111).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 80°C to 84°C
Color White
Density 0.779 g/mL
Boiling Point 150°C (0.001 mmHg)
Linear Formula CH3(CH2)38CH3
Quantity 0.5 g
Beilstein 1800152
Solubility Information Soluble in chloroform. Slightly soluble in methanol.Insoluble in water.
Formula Weight 563.09
Percent Purity ≤97%
Physical Form Flakes
Chemical Name or Material n-Tetracontane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'