missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-tert-Butylglycine hydrochloride, 97%

Brand:  Alfa Aesar™ H64270.14

Code : NEW

Additional Details : CAS Number : 6939-23-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15431389

  • £217.40 / 25g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-tert-Butylglycine hydrochloride
(CH3)3CNHCH2COOH·HCl
MFCD03788441
2-(tert-Butylamino)acetic acid hydrochloride; Intermediate of Tigecycline
6939-23-7
167.63
6296616

Specifications

N-tert-Butylglycine hydrochloride
Solid
(CH3)3CNHCH2COOH·HCl
MFCD03788441
2-(tert-Butylamino)acetic acid hydrochloride; Intermediate of Tigecycline
223° to 224°C
6939-23-7
97%
167.63
6296616
White
25g
Videos
SDS