missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-tert-Butyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65281.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409536

  • £66.46 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

N-tert-Butyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide
231.34
MFCD27975745

Specifications

N-tert-Butyl-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
231.34
1g
95%
MFCD27975745
Videos
SDS