missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-tert-Butyl-4-chloro-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65847.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15452649

  • £79.71 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-tert-Butyl-4-chloro-3-sulfamoylbenzamide
290.77
MFCD21102418

Specifications

N-tert-Butyl-4-chloro-3-sulfamoylbenzamide
Solid
290.77
1g
95%
MFCD21102418
Videos
SDS